MAT238925500 Matrix Scientific

Methyl 2-amino-5-(4-cyanophenyl)-1,3-thiazole-4-carboxylate

  • MDL number MFCD06200923
  • Cas Number : 886361-36-0
  • Molecular Weight : 259.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 266-268
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043149.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043149.html
Availability details for
Quantity
  • Availability
  • Add to Cart