MAT238925534 Matrix Scientific

Ethyl 3-(trifluoromethyl)-2-pyridinecarboxylate

  • MDL number MFCD09864691
  • Cas Number : 952182-76-2
  • Molecular Weight : 219.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043183.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043183.html
Availability details for
Quantity
  • Availability
  • Add to Cart