MAT238925536 Matrix Scientific

3-(3-Bromophenyl)-3-oxopropanenitrile

  • MDL number MFCD02260778
  • Cas Number : 70591-86-5
  • Molecular Weight : 224.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-95
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043185.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043185.html
Availability details for
Quantity
  • Availability
  • Add to Cart