MAT238925548 Matrix Scientific

1-Phenyl-1H-1,2,4-triazol-3-ol

  • MDL number MFCD00138713
  • Cas Number : 4231-68-9
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 277-280
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043197.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043197.html
Availability details for
Quantity
  • Availability
  • Add to Cart