MAT238925554 Matrix Scientific

1-[(Aminooxy)methyl]-4-methylbenzene hydrochloride

  • MDL number MFCD01114570
  • Cas Number : 38936-62-8
  • Molecular Weight : 173.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-230
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043203.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043203.html
Availability details for
Quantity
  • Availability
  • Add to Cart