MAT238925556 Matrix Scientific

2-[(Aminooxy)methyl]-1-chloro-3-fluorobenzenehydrochloride

  • MDL number MFCD01114572
  • Cas Number : 93081-15-3
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-188
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043205.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043205.html
Availability details for
Quantity
  • Availability
  • Add to Cart