MAT238925567 Matrix Scientific

1-(4-Chlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD00173066
  • Cas Number : 103058-60-2
  • Molecular Weight : 237.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-194
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043216.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043216.html
Availability details for
Quantity
  • Availability
  • Add to Cart