MAT238925573 Matrix Scientific

4-(4-Methoxyphenyl)-3-buten-2-one

  • MDL number MFCD00017251
  • Cas Number : 943-88-4
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-77
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043222.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043222.html
Availability details for
Quantity
  • Availability
  • Add to Cart