MAT238925580 Matrix Scientific

2-[(4-Chlorophenyl)sulfanyl]-2-methylpropanal

  • MDL number MFCD00243426
  • Cas Number : 56421-90-0
  • Molecular Weight : 214.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Boiling point 112�5mm
Product Link http://www.matrixscientific.com/043229.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043229.html
Availability details for
Quantity
  • Availability
  • Add to Cart