MAT238925587 Matrix Scientific

5-Amino-1-(tert-butyl)-3-(4-methylphenyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD06200927
  • Cas Number : 186896-24-2
  • Molecular Weight : 254.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-159
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/043236.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043236.html
Availability details for
Quantity
  • Availability
  • Add to Cart