MAT238925596 Matrix Scientific

Methyl 5-chloro-4-formyl-1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD01109226
  • Cas Number : 401571-11-7
  • Molecular Weight : 264.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-126
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043245.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043245.html
Availability details for
Quantity
  • Availability
  • Add to Cart