MAT238925615 Matrix Scientific

4-(2-Hydroxyethoxy)benzenecarboxylic acid

  • MDL number MFCD00020367
  • Cas Number : 1711-24-6
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-179
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043264.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043264.html
Availability details for
Quantity
  • Availability
  • Add to Cart