MAT238925626 Matrix Scientific

3-(tert-Butoxy)benzenecarboxylic acid

  • MDL number MFCD03412397
  • Cas Number : 15360-02-8
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043275.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043275.html
Availability details for
Quantity
  • Availability
  • Add to Cart