MAT238925648 Matrix Scientific

5-(4-Methylpiperazino)-2-nitroaniline

  • MDL number MFCD00545964
  • Cas Number : 23491-48-7
  • Molecular Weight : 236.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-155
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043297.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043297.html
Availability details for
Quantity
  • Availability
  • Add to Cart