MAT238925668 Matrix Scientific

3-(2-Chlorophenyl)-1H-pyrazole

  • MDL number MFCD00111882
  • Cas Number : 59843-55-9
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-77
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/043317.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043317.html
Availability details for
Quantity
  • Availability
  • Add to Cart