MAT238925670 Matrix Scientific

N-(2-Chlorophenyl)-1-hydrazinecarboxamide

  • MDL number MFCD03791165
  • Cas Number : 35580-76-8
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-116
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/043319.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043319.html
Availability details for
Quantity
  • Availability
  • Add to Cart