MAT238925680 Matrix Scientific

4-(2-Chloroacetyl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD04973717
  • Cas Number : 115027-23-1
  • Molecular Weight : 171.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-181
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043329.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043329.html
Availability details for
Quantity
  • Availability
  • Add to Cart