MAT238925766 Matrix Scientific

4-(4-Chlorophenoxy)-3-nitrobenzenecarbaldehyde

  • MDL number MFCD01113126
  • Cas Number : 320416-35-1
  • Molecular Weight : 277.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-119
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043415.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043415.html
Availability details for
Quantity
  • Availability
  • Add to Cart