MAT238925773 Matrix Scientific

4-(4-Methylphenoxy)-3-nitrobenzenecarbaldehyde

  • MDL number MFCD06659537
  • Cas Number : 750641-06-6
  • Molecular Weight : 257.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-109
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043422.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043422.html
Availability details for
Quantity
  • Availability
  • Add to Cart