MAT238925796 Matrix Scientific

2-Bromo-N-(2,6-dimethylphenyl)acetamide

  • MDL number MFCD00215011
  • Cas Number : 40251-98-7
  • Molecular Weight : 242.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-151
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043445.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043445.html
Availability details for
Quantity
  • Availability
  • Add to Cart