MAT238925804 Matrix Scientific

Methyl 2-amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c] pyridine

  • MDL number MFCD00180791
  • Cas Number : 321430-29-9
  • Molecular Weight : 391.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-151
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043453.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043453.html
Availability details for
Quantity
  • Availability
  • Add to Cart