MAT238925827 Matrix Scientific

4-Chloro-5-methoxy-2-(2-pyridinyl)pyrimidine

  • MDL number MFCD00215412
  • Cas Number : 321432-82-0
  • Molecular Weight : 221.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168-171
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043476.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043476.html
Availability details for
Quantity
  • Availability
  • Add to Cart