MAT238925834 Matrix Scientific

1-Methyl-3-phenyl-1H-pyrazol-5-ol

  • MDL number MFCD03001245
  • Cas Number : 34347-81-4
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-215
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043483.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043483.html
Availability details for
Quantity
  • Availability
  • Add to Cart