MAT238925871 Matrix Scientific

4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-benzenecarbaldehyde

  • MDL number MFCD01568788
  • Cas Number : 338393-48-9
  • Molecular Weight : 253.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-108
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043520.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043520.html
Availability details for
Quantity
  • Availability
  • Add to Cart