MAT238925872 Matrix Scientific

2-(3,4-Difluorophenoxy)benzenecarbaldehyde

  • MDL number MFCD01568793
  • Cas Number : 289717-89-1
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-64
Purity 95
Country of Origin USA
Boiling point 110-112�2mm
Product Link http://www.matrixscientific.com/043521.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043521.html
Availability details for
Quantity
  • Availability
  • Add to Cart