MAT238925916 Matrix Scientific

2,4-Difluoro-5-nitrobenzenol

  • MDL number MFCD08443942
  • Cas Number : 113512-57-5
  • Molecular Weight : 175.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043565.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043565.html
Availability details for
Quantity
  • Availability
  • Add to Cart