MAT238925917 Matrix Scientific

5-Amino-2,4-difluorobenzenol

  • MDL number MFCD04973718
  • Cas Number : 113512-71-3
  • Molecular Weight : 145.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189-190
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043566.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043566.html
Availability details for
Quantity
  • Availability
  • Add to Cart