MAT238925920 Matrix Scientific

Ethyl 2-(4-chlorobenzyl)-1,3-thiazole-4-carboxylate

  • MDL number MFCD02102512
  • Cas Number : 477872-92-7
  • Molecular Weight : 281.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-113
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043569.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043569.html
Availability details for
Quantity
  • Availability
  • Add to Cart