MAT238925934 Matrix Scientific

5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde

  • MDL number MFCD03425728
  • Cas Number : 207290-72-0
  • Molecular Weight : 211.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-139
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043583.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043583.html
Availability details for
Quantity
  • Availability
  • Add to Cart