MAT238925975 Matrix Scientific

2-(Chloromethyl)-1,3-benzothiazole

  • MDL number MFCD05664944
  • Cas Number : 37859-43-1
  • Molecular Weight : 183.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Boiling point 102�5mm
Product Link http://www.matrixscientific.com/043624.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/043624.html
Availability details for
Quantity
  • Availability
  • Add to Cart