MAT238925978 Matrix Scientific

2-Furamide

  • MDL number MFCD00236147
  • Cas Number : 609-38-1
  • Molecular Weight : 111.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-140
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043627.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043627.html
Availability details for
Quantity
  • Availability
  • Add to Cart