MAT238926007 Matrix Scientific

5-Chloro-2-oxo-1,2-dihydro-3-pyridinecarboxylic acid

  • MDL number MFCD03425803
  • Cas Number : 38076-80-1
  • Molecular Weight : 173.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 263-265
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043657.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043657.html
Availability details for
Quantity
  • Availability
  • Add to Cart