MAT238926008 Matrix Scientific

Methyl 5-chloro-6-oxo-1,6-dihydro-3-pyridinecarboxylate

  • MDL number MFCD03844853
  • Cas Number : 316166-47-9
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-222
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043658.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043658.html
Availability details for
Quantity
  • Availability
  • Add to Cart