MAT238926051 Matrix Scientific

2-Methyl-5-nitro-1,3-benzoxazole

  • MDL number MFCD00466748
  • Cas Number : 32046-51-8
  • Molecular Weight : 178.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-158
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043701.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043701.html
Availability details for
Quantity
  • Availability
  • Add to Cart