MAT238926080 Matrix Scientific

5-(4-Methoxyphenyl)-2-thiophenecarbohydrazide

  • MDL number MFCD06200930
  • Cas Number : 69202-24-0
  • Molecular Weight : 248.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-198
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/043730.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043730.html
Availability details for
Quantity
  • Availability
  • Add to Cart