MAT238926085 Matrix Scientific

4'-Nitro[1,1'-biphenyl]-4-carbaldehyde

  • MDL number MFCD06200931
  • Cas Number : 98648-23-8
  • Molecular Weight : 227.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-120
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043735.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043735.html
Availability details for
Quantity
  • Availability
  • Add to Cart