MAT238926112 Matrix Scientific

Ethyl 2-chloro-2-[2-(4-methylphenyl)hydrazono]-acetate

  • MDL number MFCD02065467
  • Cas Number : 27171-88-6
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-105
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043762.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043762.html
Availability details for
Quantity
  • Availability
  • Add to Cart