MAT238926119 Matrix Scientific

2-(2-Quinoxalinylsulfanyl)acetic acid

  • MDL number MFCD00138691
  • Cas Number : 69001-90-7
  • Molecular Weight : 220.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157-159
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043769.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043769.html
Availability details for
Quantity
  • Availability
  • Add to Cart