MAT238926134 Matrix Scientific

3-(6-Chloro-3-pyridazinyl)-1H-indole

  • MDL number MFCD00214721
  • Cas Number : 129287-26-9
  • Molecular Weight : 229.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 270-272
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043784.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043784.html
Availability details for
Quantity
  • Availability
  • Add to Cart