MAT238926160 Matrix Scientific

Ethyl 2-(2-chlorophenyl)-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD01935973
  • Cas Number : 338982-17-5
  • Molecular Weight : 281.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-105
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043810.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043810.html
Availability details for
Quantity
  • Availability
  • Add to Cart