MAT238926188 Matrix Scientific

2-Amino-4-methoxynicotinonitrile

  • MDL number MFCD00975441
  • Cas Number : 98651-70-8
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043838.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043838.html
Availability details for
Quantity
  • Availability
  • Add to Cart