MAT238926189 Matrix Scientific

4-Methoxy-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

  • MDL number MFCD00975442
  • Cas Number : 21642-98-8
  • Molecular Weight : 150.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 293
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043839.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043839.html
Availability details for
Quantity
  • Availability
  • Add to Cart