MAT238926192 Matrix Scientific

5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00244899
  • Cas Number : 883-38-5
  • Molecular Weight : 220.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-64
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043842.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043842.html
Availability details for
Quantity
  • Availability
  • Add to Cart