MAT238926200 Matrix Scientific

2-[(4-Methylphenyl)sulfanyl]nicotinaldehyde

  • MDL number MFCD00794313
  • Cas Number : 338982-20-0
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-84
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043850.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043850.html
Availability details for
Quantity
  • Availability
  • Add to Cart