MAT238926226 Matrix Scientific

2-[3-(Trifluoromethyl)phenoxy]benzenecarbaldehyde

  • MDL number MFCD01814776
  • Cas Number : 408369-24-4
  • Molecular Weight : 266.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Boiling point 108-110�1mm
Product Link http://www.matrixscientific.com/043876.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043876.html
Availability details for
Quantity
  • Availability
  • Add to Cart