MAT238926245 Matrix Scientific

1-[4-(3-Fluoropropoxy)phenyl]-1-ethanone

  • MDL number MFCD03001251
  • Cas Number : 400878-19-5
  • Molecular Weight : 196.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043895.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043895.html
Availability details for
Quantity
  • Availability
  • Add to Cart