MAT238926271 Matrix Scientific

1-(3,4-Dichlorobenzyl)-4-piperidine-carboxylic acid hydrochloride

  • MDL number MFCD03425739
  • Cas Number : 451485-54-4
  • Molecular Weight : 324.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260-262
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043921.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043921.html
Availability details for
Quantity
  • Availability
  • Add to Cart