MAT238926288 Matrix Scientific

4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one

  • MDL number MFCD03617955
  • Cas Number : 258281-02-6
  • Molecular Weight : 156.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-81
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043938.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043938.html
Availability details for
Quantity
  • Availability
  • Add to Cart