MAT238926292 Matrix Scientific

7-Amino-2-methyl-3-[3-(trifluoromethyl)phenyl]-pyrazolo[1,5-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD03791193
  • Cas Number : 863305-82-2
  • Molecular Weight : 336.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224-226
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043942.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043942.html
Availability details for
Quantity
  • Availability
  • Add to Cart