MAT238926305 Matrix Scientific

2-Ethyl-1,3-benzoxazol-5-amine

  • MDL number MFCD03791201
  • Cas Number : 204771-75-5
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-77
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043955.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043955.html
Availability details for
Quantity
  • Availability
  • Add to Cart